ADVH49B837DC AChemBlock

(1R)-5-METHOXY-1,2,3,4-TETRAHYDRONAPHTHALEN-1-AMINE

  • Cas Number : 103882-27-5
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1r-5-methoxy-1-2-3-4-tetrahydronaphthalen-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89765
Availability details for
Quantity
  • Availability
  • Add to Cart