ADVH49B837DD AChemBlock

N-DESMETHYLZOPICLONE HYDROCHLORIDE

  • Cas Number : 1216659-29-8
  • Molecular Weight : 411.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-desmethylzopiclone-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89766
Availability details for
Quantity
  • Availability
  • Add to Cart