ADVH49B837E0 AChemBlock

NNC-0640

  • Cas Number : 307986-98-7
  • Molecular Weight : 573.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/nnc-0640.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89771
Availability details for
Quantity
  • Availability
  • Add to Cart