ADVH49B837E2 AChemBlock

3-pyridine toxoflavin

  • Cas Number : 32502-20-8
  • Molecular Weight : 270.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-pyridine-toxoflavin.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89773
Availability details for
Quantity
  • Availability
  • Add to Cart