ADVH49B837E3 AChemBlock

IRAK4-IN-3

  • Cas Number : 2196204-23-4
  • Molecular Weight : 385.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/irak4-in-3.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89774
Availability details for
Quantity
  • Availability
  • Add to Cart