ADVH49B837E4 AChemBlock

MW-150

  • Cas Number : 1628502-91-9
  • Molecular Weight : 381.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/mw-150.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89778
Availability details for
Quantity
  • Availability
  • Add to Cart