ADVH49B837E7 AChemBlock

Radiprodil

  • Cas Number : 496054-87-6
  • Molecular Weight : 397.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/radiprodil.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89784
Availability details for
Quantity
  • Availability
  • Add to Cart