ADVH49B837E8 AChemBlock

RU320521

  • Cas Number : 2262452-06-0
  • Molecular Weight : 415.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ru320521.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89785
Availability details for
Quantity
  • Availability
  • Add to Cart