ADVH49B837EA AChemBlock

DMX-5084

  • Cas Number : 2306178-56-1
  • Molecular Weight : 361.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/dmx-5084.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89791
Availability details for
Quantity
  • Availability
  • Add to Cart