ADVH49B837EB AChemBlock

tert-butyl 3,3-difluoro-4-hydroxy-5-(hydroxymethyl)piperidine-1-carboxylate Cis-Trans mixture

  • Cas Number : 1936599-97-1
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-3-difluoro-4-hydroxy-5-hydroxymethyl-piperidine-1-carboxylate-cis-trans-mixture.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89792
Availability details for
Quantity
  • Availability
  • Add to Cart