ADVH49B837ED AChemBlock

BI-4464

  • Cas Number : 1227948-02-8
  • Molecular Weight : 555.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bi-4464.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89797
Availability details for
Quantity
  • Availability
  • Add to Cart