ADVH49B837EE AChemBlock

OSU-T 315

  • Cas Number : 1333146-24-9
  • Molecular Weight : 533.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/osu-t-315.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89798
Availability details for
Quantity
  • Availability
  • Add to Cart