ADVH49B837F0 AChemBlock

AMG-333

  • Cas Number : 1416799-28-4
  • Molecular Weight : 453.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/amg-333.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89800
Availability details for
Quantity
  • Availability
  • Add to Cart