ADVH49B837F1 AChemBlock

GSK467

  • Cas Number : 1628332-52-4
  • Molecular Weight : 319.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/gsk467.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89801
Availability details for
Quantity
  • Availability
  • Add to Cart