ADVH49B837F3 AChemBlock

HS-10296

  • Cas Number : 1899921-05-1
  • Molecular Weight : 525.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/hs-10296.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89803
Availability details for
Quantity
  • Availability
  • Add to Cart