ADVH49B837F5 AChemBlock

(Cis)-tert-butyl 2,4-difluoro-3,3-dihydroxy-8-azabicyclo[3 2 1]octane-8-carboxylate racemate

  • Molecular Weight : 279.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/cis-tert-butyl-2-4-difluoro-3-3-dihydroxy-8-azabicyclo-3-2-1-octane-8-carboxylate-racemate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89809
Availability details for
Quantity
  • Availability
  • Add to Cart