ADVH7F37D5C3 AChemBlock

3-Aminoazetidine-3-carboxylic acid

  • MDL number MFCD12406945
  • Cas Number : 138650-25-6
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form off-white powder
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-aminoazetidine-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3611
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart