ADVH7F37D5FE AChemBlock

3-(Hydroxymethyl)-3-methylpiperidine hydrochloride

  • MDL number MFCD22421713
  • Cas Number : 955027-74-4
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form white solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxymethyl-3-methylpiperidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2935
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart