ADVH7F37D6A7 AChemBlock

8-Boc-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene

  • MDL number MFCD22380006
  • Cas Number : 185099-68-7
  • Molecular Weight : 357.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-boc-3-trifluoromethylsulfonyloxy-8-azabicyclo-3-2-1-oct-3-ene.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4150
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart