ADVH7F37D6C3 AChemBlock

8-Cbz-2-oxo-3-oxa-1,8-diaza-spiro[5.5]undecane

  • MDL number MFCD23105916
  • Cas Number : 1408074-70-3
  • Molecular Weight : 304.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-cbz-2-oxo-3-oxa-1-8-diaza-spiro-5-5-undecane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5083
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart