ADVH7F37D800 AChemBlock

1-Boc-3-mercaptopiperidine

  • MDL number MFCD10566038
  • Cas Number : 1017798-34-3
  • Molecular Weight : 217.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form orange liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-mercaptopiperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6616
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart