ADVH7F37D8CC AChemBlock

5-Hydroxy-1-Boc-hexahydrocyclopenta[b]pyrrole

  • MDL number MFCD23106464
  • Cas Number : 1419101-49-7
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-hydroxy-1-boc-hexahydrocyclopenta-b-pyrrole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6884
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart