ADVH7F37D91E AChemBlock

(1R,6R)-rel-3-Boc-5,5-dimethyl-3,7-diazabicyclo[4.2.0]octane

  • MDL number MFCD17016508
  • Cas Number : 1251021-71-2
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1r-6r-rel-3-boc-5-5-dimethyl-3-7-diazabicyclo-4-2-0-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7232
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart