ADVH7F37D927 AChemBlock

4,7-Dihydro-imidazole[4,5-d]1,3-diazepine-8(1H)-one hydrochloride

  • MDL number MFCD23105840
  • Cas Number : 71222-44-1
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-7-dihydro-imidazole-4-5-d-1-3-diazepine-8-1h-one-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7279
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart