ADVH7F37D9AD AChemBlock

1-Benzyl-3-fluoropiperidin-4-one

  • MDL number MFCD11847801
  • Cas Number : 475152-19-3
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-3-fluoropiperidin-4-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8149
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart