ADVH7F37D9C6 AChemBlock

4,5-Dihydro-6-oxime-imidazo[4,5,1-jk][1]benzazepine-2,6,7(1H)-trione

  • MDL number MFCD26793907
  • Cas Number : 92260-82-7
  • Molecular Weight : 231.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-5-dihydro-6-oxime-imidazo-4-5-1-jk-1-benzazepine-2-6-7-1h-trione.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7656
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart