ADVH7F37D9D7 AChemBlock

(5-Bromo-2-pyridyl)methanamine hydrochloride

  • MDL number MFCD20923716
  • Cas Number : 1241911-26-1
  • Molecular Weight : 223.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-pyridyl-methanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7736
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart