ADVH7F37D9D8 AChemBlock

2-(2-Methoxyethyl)pyrrolidine

  • MDL number MFCD19102570
  • Cas Number : 933746-73-7
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-methoxyethyl-pyrrolidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7743
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart