ADVH7F37D9DA AChemBlock

6,7-Dihydro-1-methyl-1H-indol-4(5H)-one

  • MDL number MFCD01444760
  • Cas Number : 51471-08-0
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-7-dihydro-1-methyl-1h-indol-4-5h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7745
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart