ADVH7F37D9FB AChemBlock

(1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxylic acid trifluoroacetate

  • MDL number MFCD27987984
  • Cas Number : 1523541-80-1
  • Molecular Weight : 241.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1s-3s-5s-2-azabicyclo-3-1-0-hexane-3-carboxylic-acid-trifluoroacetate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7862
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart