ADVH7F37DA2C AChemBlock

4-Amino-6-methyl-pyrimidine-2-carboxylic acid methyl ester

  • MDL number MFCD21741927
  • Cas Number : 1520279-43-9
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-amino-6-methyl-pyrimidine-2-carboxylic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9132
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart