ADVH7F37DC6B AChemBlock

Methyl 3-iodo-1H-indole-7-carboxylate

  • MDL number MFCD28501544
  • Cas Number : 1788043-93-5
  • Molecular Weight : 301.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/methyl-3-iodo-1h-indole-7-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=L13620
pictograms GHS06
hstatements H301;H319;H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart