ADVH7F37DD87 AChemBlock

1-[1-(Propan-2-yl)piperidin-4-yl]ethan-1-ol

  • MDL number MFCD30146457
  • Cas Number : 1699407-61-8
  • Molecular Weight : 171.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/1-1-propan-2-yl-piperidin-4-yl-ethan-1-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=L14280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart