ADVH7F37DEAA AChemBlock

2-chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD27987389
  • Cas Number : 1638767-99-3
  • Molecular Weight : 211.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/2-chloro-7-methyl-7h-pyrrolo-2-3-d-pyrimidine-6-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=L14852
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart