ADVH7F37E119 AChemBlock

Trans-3-fluoro-3-methylcyclobutane-1-carboxylic acid

  • MDL number MFCD28969665
  • Cas Number : 1455037-43-0
  • Molecular Weight : 132.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/trans-3-fluoro-3-methylcyclobutane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=L21324
pictograms GHS07
hstatements H303;H320
Availability details for
Quantity
  • Availability
  • Add to Cart