ADVH7F37E1ED AChemBlock

(S)-Methyl 2-amino-2-(4-fluorophenyl)acetate hydrochloride

  • MDL number MFCD12910960
  • Cas Number : 916602-09-0
  • Molecular Weight : 219.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/s-methyl-2-amino-2-4-fluorophenyl-acetate-hydrochloride.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=M22709
pictograms GHS07
hstatements H303;H320
Availability details for
Quantity
  • Availability
  • Add to Cart