ADVH7F37E318 AChemBlock

5-Bromo-D-tryptophan

  • MDL number MFCD09264327
  • Cas Number : 93299-40-2
  • Molecular Weight : 283.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/5-bromo-d-tryptophan.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=M22661
pictograms GHS07
hstatements H303;H320
Availability details for
Quantity
  • Availability
  • Add to Cart