ADVH7F37F52F AChemBlock

(R)-3-Amino-3-(4-methoxyphenyl)propionic acid

  • MDL number MFCD01318245
  • Cas Number : 131690-57-8
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/r-3-amino-3-4-methoxyphenyl-propionic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=O28958
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart