ADVH7F37F639 AChemBlock

1-(4-(trifluoromethoxy)benzyl)-5-methyl-1H-1,2,3-triazol-4-amine

  • MDL number MFCD22703232
  • Cas Number : 1511177-48-2
  • Molecular Weight : 272.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-4-trifluoromethoxy-benzyl-5-methyl-1h-1-2-3-triazol-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29253
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart