ADVH7F37F63B AChemBlock

1,3-dichloro-5-(3-methylbut-1-en-2-yl)benzene

  • MDL number MFCD28656718
  • Cas Number : 864736-69-6
  • Molecular Weight : 215.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-dichloro-5-3-methylbut-1-en-2-yl-benzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29260
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart