ADVH7F37F63E AChemBlock

Imidodicarbonic acid,2-(6-bromo-1-isoquinolinyl)-,1,3-bis(1,1-dimethylethyl) ester

  • MDL number MFCD27924172
  • Cas Number : 911305-48-1
  • Molecular Weight : 423.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/imidodicarbonic-acid-2-6-bromo-1-isoquinolinyl-1-3-bis-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29267
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart