ADVH7F37F641 AChemBlock

methyl 4-(3-aminopropyl)benzoate hydrochloride

  • MDL number MFCD09865097
  • Cas Number : 74733-37-2
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-3-aminopropyl-benzoate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29273
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart