ADVH7F37F644 AChemBlock

4-methoxy-3-(methylthio)benzoic acid

  • MDL number MFCD20640312
  • Cas Number : 87346-53-0
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-3-methylthio-benzoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29289
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart