ADVH7F37F646 AChemBlock

2-(chloromethyl)-2H-1,2,3-triazole

  • MDL number MFCD19235522
  • Cas Number : 80199-90-2
  • Molecular Weight : 117.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloromethyl-2h-1-2-3-triazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29307
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart