ADVH7F37F647 AChemBlock

2,6-bis(methylthio)benzaldehyde

  • MDL number MFCD22689676
  • Cas Number : 918882-53-8
  • Molecular Weight : 198.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-6-bis-methylthio-benzaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29309
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart