ADVH7F37F652 AChemBlock

7-methoxy-5-methylbenzo[b]thiophene-3-carbaldehyde

  • MDL number MFCD22560627
  • Cas Number : 1388061-05-9
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-methoxy-5-methylbenzo-b-thiophene-3-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29380
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart