ADVH7F37F654 AChemBlock

5-bromo-3-(difluoromethoxy)-2-methoxypyridine

  • MDL number MFCD21605094
  • Cas Number : 1241752-50-0
  • Molecular Weight : 254.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-difluoromethoxy-2-methoxypyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29385
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart