ADVH7F37F663 AChemBlock

2-(4-fluorophenyl)pyridin-4-amine

  • MDL number MFCD06739106
  • Cas Number : 886366-09-2
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-fluorophenyl-pyridin-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29444
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart