ADVH7F37F664 AChemBlock

ethyl 2-(4-methoxyphenyl)cyclopropanecarboxylate

  • MDL number MFCD02258631
  • Cas Number : 98017-60-8
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-4-methoxyphenyl-cyclopropanecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29447
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart