ADVH7F37F66C AChemBlock

imidazo[1,2-a]pyrazin-3-amine hydrochloride

  • MDL number MFCD27924163
  • Cas Number : 56888-69-8
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-2-a-pyrazin-3-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29476
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart